Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PADI4 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB337739 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337739 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337739 Supplier Novus Biologicals Supplier No. NBP197309MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PADI4 Polyclonal antibody specifically detects PADI4 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot (Negative), ICC/IF (Negative)

Specifications

Antigen PADI4
Applications Immunohistochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC/IF (Negative)
Formulation 50mM Sodium Borate
Gene Alias HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PAD4 protein (between residues 200-250) [UniProt Q9UM07]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23569
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.