Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PADI4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP238499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP238499 0.1 mL
NB436189 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP238499 Supplier Novus Biologicals Supplier No. NBP238499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PADI4 Polyclonal specifically detects PADI4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PADI4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9UM07
Gene Alias HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4
Gene Symbols PADI4
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: GTLTQLDICSSAPEDCTSFSINASPGVVVDIAHGPPAKKKSTGSSTWPLDPGVEVTLTMKVASGSTGDQKVQISYYGPKTPPVKALLYLTGV
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23569
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.