Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PADI4 Antibody (OTI5C10), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP273228A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273228A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273228A70 Supplier Novus Biologicals Supplier No. NBP273228AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PADI4 Monoclonal specifically detects PADI4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PADI4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5C10
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4
Gene Symbols PADI4
Host Species Mouse
Immunogen Full length recombinant protein of human PADI4 (NP_036519) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23569.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.