Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAFAH1B2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1563020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1563020 20 μL
NBP156301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1563020 Supplier Novus Biologicals Supplier No. NBP15630120UL

Rabbit Polyclonal Antibody

PAFAH1B2 Polyclonal specifically detects PAFAH1B2 in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen PAFAH1B2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O35264
Gene Alias EC 3.1.1.47, intracellular platelet-activating factor acetylhydrolase alpha 2 subunit, PAF acetylhydrolase 30 kDa subunit, PAF-AH 30 kDa subunit, PAFAH subunit beta, PAF-AH subunit beta, PAF-AH1b alpha 2 subunit, PAFAHB, platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa), platelet-activating factor acetylhydrolase IB subunit beta, platelet-activating factor acetylhydrolase, isoform Ib, beta subunit 30kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa)
Gene Symbols PAFAH1B2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PAFAH1B2(platelet-activating factor acetylhydrolase, isoform Ib, beta subunit 30kDa) The peptide sequence was selected from the N terminal of PAFAH1B2. Peptide sequence MSQGDSNPAAIPHAAEDIQGDDRWMSQHNRFVLD
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5049
Test Specificity This product is specific to Subunit or Isoform: beta.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.