Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAFAH1B2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PAFAH1B2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1563020
![]() |
Novus Biologicals
NBP15630120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156301
![]() |
Novus Biologicals
NBP156301 |
100 μL |
Each for $487.50
|
|
|||||
Description
PAFAH1B2 Polyclonal specifically detects PAFAH1B2 in Human samples. It is validated for Western Blot.Specifications
PAFAH1B2 | |
Polyclonal | |
Rabbit | |
O35264 | |
5049 | |
IgG | |
This product is specific to Subunit or Isoform: beta. |
Western Blot | |
Unconjugated | |
RUO | |
EC 3.1.1.47, intracellular platelet-activating factor acetylhydrolase alpha 2 subunit, PAF acetylhydrolase 30 kDa subunit, PAF-AH 30 kDa subunit, PAFAH subunit beta, PAF-AH subunit beta, PAF-AH1b alpha 2 subunit, PAFAHB, platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa), platelet-activating factor acetylhydrolase IB subunit beta, platelet-activating factor acetylhydrolase, isoform Ib, beta subunit 30kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa) | |
PAFAH1B2 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title