Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAFAH1B3 Antibody (OTI5D2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N273229A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273229A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273229A405 Supplier Novus Biologicals Supplier No. NBP273229AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAFAH1B3 Monoclonal specifically detects PAFAH1B3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PAFAH1B3
Applications Western Blot
Classification Monoclonal
Clone OTI5D2
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.1.1.47, FLJ44990, PAF acetylhydrolase 29 kDa subunit, PAF-AH 29 kDa subunit, PAFAH subunit gamma, PAF-AH subunit gamma, PAF-AH1b alpha 1 subunit, PAFAHG, platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa), platelet-activating factor acetylhydrolase IB subunit gamma, platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit (29kD), platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa)
Gene Symbols PAFAH1B3
Host Species Mouse
Immunogen Full length recombinant protein of human PAFAH1B3 (NP_001139411) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5050.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.