Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAG1 Antibody (MEM-255), PerCP, Novus Biologicals™
SDP

Catalog No. NB500342PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500342PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500342PCP Supplier Novus Biologicals Supplier No. NB500342PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAG1 Monoclonal specifically detects PAG1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PAG1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MEM-255
Conjugate PerCP
Dilution Flow Cytometry
Gene Accession No. Q9NWQ8
Gene Alias CBPMGC138364, Csk-binding protein, PAGFLJ37858, phosphoprotein associated with glycosphingolipid microdomains 1, phosphoprotein associated with glycosphingolipid-enriched microdomains 1, Transmembrane adapter protein PAG, Transmembrane phosphoprotein Cbp
Gene Symbols PAG1
Host Species Mouse
Immunogen Recombinant intracellular fragment, corresponding to amino acids 97-432 of Human Cbp/PAG (NM_018440.3).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 55824
Test Specificity The antibody MEM-255 recognizes an epitope (aa 235-280) of Csk-binding protein (Cbp) located in the cytoplasmic domain, also known as protein associated with glycosphingolipid-enriched microdomains (PAG).
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.