Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAG1 Antibody (PAG-C1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB500487IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500487IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500487IR Supplier Novus Biologicals Supplier No. NB500487IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAG1 Monoclonal specifically detects PAG1 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PAG1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PAG-C1
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CBPMGC138364, Csk-binding protein, PAGFLJ37858, phosphoprotein associated with glycosphingolipid microdomains 1, phosphoprotein associated with glycosphingolipid-enriched microdomains 1, Transmembrane adapter protein PAG, Transmembrane phosphoprotein Cbp
Gene Symbols PAG1
Host Species Mouse
Immunogen C-terminal peptide (last 15 amino acids) of human Csk binding protein coupled to KLH.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 55824
Test Specificity The antibody PAG-C1 recognizes an epitope located in the C-terminal domain of Csk-binding protein (Cbp PAG), a 46 kDa ubiquitously expressed transmembrane adaptor protein present in membrane rafts (glycosphingolipid-enriched microdomains), which however migrates on SDS PAGE gels anomalously as an 80 kDa molecule.
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.