Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAIP2 Antibody (1C12), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB310039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310039 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310039 Supplier Novus Biologicals Supplier No. NBP222555MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAIP2 Monoclonal antibody specifically detects PAIP2 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen PAIP2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C12
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias MGC72018, PABC1-interacting protein 2, PABP-interacting protein 2, PAIP-2, PAIP2APoly(A)-binding protein-interacting protein 2, poly(A) binding protein interacting protein 2, polyA-binding protein-interacting protein 2, polyadenylate-binding protein-interacting protein 2
Host Species Mouse
Immunogen Recombinant human PAIP2 (1-127aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51247
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.