Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK1/2/3, p Thr423, p Thr402, p Thr421 Antibody, Novus Biologicals™
SDP

Catalog No. NB10082131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10082131 0.05 mL
NB017873 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10082131 Supplier Novus Biologicals Supplier No. NB10082131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

PAK1/2/3 (p Thr423, p Thr402, p Thr421) Polyclonal specifically detects PAK1/2/3 (p Thr423, p Thr402, p Thr421) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen PAK1/2/3 (p Thr423, p Thr402, p Thr421)
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide
Gene Accession No. Q13153//Q13177//O75914
Gene Alias alpha-PAK, EC 2.7.11, EC 2.7.11.1, MGC130000, MGC130001, p21 protein (Cdc42/Rac)-activated kinase 1, p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast), p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related), p21-activated kinase 1, p65-PAK, PAK-1, PAKalpha, serine/threonine-protein kinase PAK 1, STE20 homolog, yeast
Gene Symbols PAK1
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from Human PAK1/2/3 around the phosphorylation site of threonine 423/402/421 (R-S-TP-M-V).
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5058
Test Specificity Detects endogenous levels of PAK1/2/3 only when phosphorylated at threonine 423/402/421.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.