Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK1, p Ser204 Antibody, Novus Biologicals™
SDP

Catalog No. NB019418 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019418 0.025 mL
NBP229665 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019418 Supplier Novus Biologicals Supplier No. NBP2296650.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAK1 Polyclonal specifically detects PAK1 in Human samples. It is validated for Western Blot.

Specifications

Antigen PAK1 (p Ser204)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 50% Glycerol with 0.05% Sodium Azide
Gene Accession No. Q13153
Gene Alias alpha-PAK, EC 2.7.11, EC 2.7.11.1, MGC130000, MGC130001, p21 protein (Cdc42/Rac)-activated kinase 1, p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast), p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related), p21-activated kinase 1, p65-PAK, PAK-1, PAKalpha, serine/threonine-protein kinase PAK 1, STE20 homolog, yeast
Gene Symbols PAK1
Host Species Rabbit
Immunogen Amino Acids encompassing and including phosphorylated Ser199 and Ser204 of human PAK1. Sequence is highly homologous (greater than 90%) to PAK2 and PAK3.
Molecular Weight of Antigen 65 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5058
Test Specificity Recognizes PAK1 phosphorylated on Ser199 and Ser204 (human)/Ser198 and Ser203 (mouse). Immunogen sequence is higly homolgous (greater than 90%) with PAK2 and PAK3.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.