Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176721 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176721 Supplier Novus Biologicals Supplier No. NBP176721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAK2 Polyclonal specifically detects PAK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PAK2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_002568
Gene Alias EC 2.7.11, gamma-PAK, p21 (CDKN1A)-activated kinase 2, p21 protein (Cdc42/Rac)-activated kinase 2, p21-activated kinase 2, p58, PAK-2, PAK65EC 2.7.11.1, PAKgamma, S6/H4 kinase, serine/threonine-protein kinase PAK 2
Gene Symbols PAK2
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human PAK2. The immunogen is located within amino acids 20 - 70 of PAK2. Amino Acid Squence: PLPSVPEEKKPRHK
Molecular Weight of Antigen 58 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5062
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.