Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK3 (p Ser144, p Ser141, p Ser139) Rabbit anti-Human, Mouse, Rat, Clone: SU03-70, Novus Biologicals™
SDP

Catalog No. NBP267484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267484 Supplier Novus Biologicals Supplier No. NBP267484

Rabbit Monoclonal Antibody

PAK3 (p Ser144, p Ser141, p Ser139) Monoclonal antibody specifically detects PAK3 (p Ser144, p Ser141, p Ser139) in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin).

Specifications

Antigen PAK3 (p Ser144, p Ser141, p Ser139)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SU03-70
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias beta-PAK, bPAK, CDKN1A, EC 2.7.11, EC 2.7.11.1, hPAK3, mental retardation, X-linked 47, MRX30, MRX47, Oligophrenin-3, OPHN3, p21 (CDKN1A)-activated kinase 3, p21 protein (Cdc42/Rac)-activated kinase 3, p21-activated kinase 3, PAK-3, PAK3beta, serine/threonine-protein kinase PAK 3
Gene Symbols PAK3
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Ser144 of Human PAK1 aa 121-150/ 545. (SwissProt: O75914 Human; SwissProt: Q13153 Human; SwissProt: Q13177 Human; SwissProt: O88643 Mouse; SwissProt: Q61036 Mouse; SwissProt: Q8CIN4 Mouse; SwissProt: P35465 Rat; SwissProt: Q62829 Rat; SwissProt: Q64303 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5063
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.