Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK4 Antibody (OTI1C7), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP273231IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273231IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273231IR Supplier Novus Biologicals Supplier No. NBP273231IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAK4 Monoclonal specifically detects PAK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PAK4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1C7
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.1, KIAA1142, p21 protein (Cdc42/Rac)-activated kinase 4, p21(CDKN1A)-activated kinase 4, p21-activated kinase 4, PAK-4, protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs, serine/threonine-protein kinase PAK 4
Gene Symbols PAK4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-308 of human PAK4 (NP_005875) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10298.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.