Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PALB2 Antibody (M41-P3F3), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250232UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250232UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250232UV Supplier Novus Biologicals Supplier No. NBP250232UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PALB2 Monoclonal specifically detects PALB2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen PALB2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone M41-P3F3
Conjugate DyLight 350
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3
Gene Symbols PALB2
Host Species Mouse
Immunogen Peptide Sequence - KENQFLMPPEE
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 79728
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.