Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladin Antibody (1E6), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP1299A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP1299A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP1299A70 Supplier Novus Biologicals Supplier No. NBP125959AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Palladin Monoclonal specifically detects Palladin in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Palladin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone 1000000
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CGI151, CGI-151, FLJ22190, FLJ38193, KIAA0992FLJ39139, MYN, myoneurin, palladin, palladin, cytoskeletal associated protein, PNCA1, Sarcoma antigen NY-SAR-77, SIH002FLJ61376
Gene Symbols PALLD
Host Species Mouse
Immunogen Endogenous Palladin immunoprecipitated from chicken embryo fibroblasts.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cellular Markers, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23022.0
Target Species Human, Mouse, Rat, Canine, Chicken
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.