Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladin Antibody (1E6), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP125959IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP125959IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP125959IR Supplier Novus Biologicals Supplier No. NBP125959IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Palladin Monoclonal specifically detects Palladin in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Palladin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone 1000000
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Gene Accession No. Q8WX93
Gene Alias CGI151, CGI-151, FLJ22190, FLJ38193, KIAA0992FLJ39139, MYN, myoneurin, palladin, palladin, cytoskeletal associated protein, PNCA1, Sarcoma antigen NY-SAR-77, SIH002FLJ61376
Gene Symbols PALLD
Host Species Mouse
Immunogen Endogenous Palladin immunoprecipitated from chicken embryo fibroblasts.
Molecular Weight of Antigen 85 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23022
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.