Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) nitrate hydrate, 99.8% (metals basis), Pd 39% min

Catalog No. AA1103518
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1103518 50 g
AA1103504 2 g
AA1103509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1103518 Supplier Thermo Scientific Chemicals Supplier No. 01103518
Only null left
Add to Cart

CAS: 207596-32-5 | N2O6Pd | 230.43 g/mol

Palladium(II) nitrate hydrate is used as a precursor to supported palladium catalysts. It is also used as a catalyst in the conversion of alkene nitration to glycol dinitrates. It is involved in the separation of chlorine and iodine and used as biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207596-32-5
Molecular Formula N2O6Pd
Molecular Weight (g/mol) 230.43
MDL Number MFCD00011169
InChI Key GPNDARIEYHPYAY-UHFFFAOYSA-N
Synonym palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate
PubChem CID 24932
IUPAC Name palladium(2+);dinitrate
SMILES [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point (decomposition)
Color Red to Brown
Physical Form Crystals
Quantity 50 g
Assay Percent Range 99.8% (Metals Basis)
Solubility Information Soluble in dilute nitric acid.
Formula Weight 230.43 (Anhydrous)
Sensitivity Hygroscopic
Appearance Red-Brown Crystals
Chemical Name or Material Palladium(II) Nitrate Hydrate
Hazard Category H272-H290-H302-H314-H335
Precautionary Statement P210-P220-P221-P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation DOT Class: 5.1; Packaging Group: II
EINECSNumber 233-265-8
RTECSNumber QV0560000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.