Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) sulfate dihydrate, Premion™, 99.95% (metals basis), Pd 44.1% min, Thermo Scientific Chemicals

Catalog No. AA1216506
Click to view available options
Quantity:
5 g

Palladium(II) sulfate dihydrate is used as a raw material to prepare other palladium complexes. It acts as a catalyst in synthetic chemistry. It is also used in electroplating. In the presence of sulfuric acid, it is used for activation of alkanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13444-98-9
Molecular Formula H4O6PdS
Molecular Weight (g/mol) 238.51
MDL Number MFCD00011173
InChI Key TWIRRPLUAGEFNJ-UHFFFAOYSA-L
Synonym palladium ii sulfate dihydrate, palladium ii sulfate dihydrate, premion, palladium sulfate dihydrate, so4.pd.2h2o, palladium 2+ sulfate dihydrate, palladium 2+ dihydrate sulfate, palladium 2+ sulfate-water 1/1/2, palladium 2+ ion dihydrate sulfate, palladium ii sulfate dihydrate metals basis
PubChem CID 22042048
IUPAC Name palladium(2+);sulfate;dihydrate
SMILES O.O.[Pd++].[O-]S([O-])(=O)=O

Specifications

Melting Point (decomposition)
Color Brown to Red
Physical Form Crystalline
Quantity 5 g
Linear Formula PdSO4·2H2O
UN Number UN3260
Solubility Information Soluble in cold water.
Formula Weight 238.51 (202.48 Anhydrous)
Percent Purity 99.95%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pd 44.1% min.
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Palladium(II) sulfate dihydrate
Show More Show Less
Hazard Category H290-H302-H314-H334-H335
Hazard Statement GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 236-957-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.