Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pallidin Antibody (OTI1H9), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB273233A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273233A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273233A48 Supplier Novus Biologicals Supplier No. NBP273233AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pallidin Monoclonal specifically detects Pallidin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Pallidin
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1H9
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias PALLID, pallid (mouse) homolog, pallidin, Pallid protein homolog, pallidin, pallidin homolog (mouse), PAsyntaxin 13 binding protein 1, Syntaxin 13-interacting protein, syntaxin 13-interacting protein pallid
Gene Symbols BLOC1S6
Host Species Mouse
Immunogen Full length recombinant protein of human Pallidin (NP_036520) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26258.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.