Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PALMD Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP276369 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP276369 Supplier Novus Biologicals Supplier No. NBP276369

Rabbit Polyclonal Antibody

PALMD Polyclonal specifically detects PALMD in Human samples. It is validated for Western Blot, Immunoprecipitation, Tissue Culture Substratum.

Specifications

Antigen PALMD
Applications Western Blot, Immunoprecipitation, Immunoassay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation 50-100 μL/mg lysate, Tissue Culture Substratum
Formulation Tris-buffered Saline, 0.1% BSA with 0.09% Sodium Azide
Gene Alias C1orf11, palmdelphin, PALMLFLJ20271, Paralemnin-like protein
Gene Symbols PALMD
Host Species Rabbit
Immunogen Immunogen sequence between amino acids 325 and 375
Purification Method Antigen Affinity-purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 54873.0
Target Species Human
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.