Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PALS1/MPP5 Antibody (OTI2D3), Novus Biologicals™
SDP

Catalog No. NBP201880 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP201880 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP201880 Supplier Novus Biologicals Supplier No. NBP201880
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PALS1/MPP5 Monoclonal specifically detects PALS1/MPP5 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen PALS1/MPP5
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI2D3
Concentration 0.88 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Flow Cytometry 1:100
Gene Alias MAGUK p55 subfamily member 5, membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5), PALS1FLJ12615, stardust
Gene Symbols MPP5
Host Species Mouse
Immunogen Full length human recombinant protein of human MPP5(NP_071919) produced in HEK293T cell.
Molecular Weight of Antigen 77.1 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64398
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.