Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan Actin Antibody (ACTN05 (C4)), Novus Biologicals™
SDP

Catalog No. NB600535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600535 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600535 Supplier Novus Biologicals Supplier No. NB600535

Mouse Monoclonal Antibody has been used in 15 publications

pan Actin Monoclonal specifically detects pan Actin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Protozoa, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen pan Actin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ACTN05 (C4)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2500 - 1:10000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 2 ug, Immunohistochemistry-Paraffin 1:50
Gene Alias ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3
Gene Symbols ACTA1
Host Species Mouse
Immunogen Chicken gizzard actin
Purification Method Protein G purified
Quantity 500 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 58
Test Specificity This antibody reacts with all six known vertebrate isoforms of actin (approximately 42kDa). It also reacts with two cytoplasmic actins (Beta, gamma) which are highly homologous to one another but which differ from the muscle actins at about 25 amino acid residues.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Protozoa, Rabbit
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.