Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pan Endothelial Cell Marker Antibody (RM0061-4J13), HRP, Novus Biologicals™
SDP

Catalog No. NBP212438H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212438H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212438H Supplier Novus Biologicals Supplier No. NBP212438H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Pan Endothelial Cell Marker Monoclonal specifically detects Pan Endothelial Cell Marker in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Pan Endothelial Cell Marker
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0061-4J13
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Host Species Rat
Immunogen Cell lysates of mouse endothelial cells
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.