Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pan-Endothelium Marker, p Ser123, p Ser124 Antibody (HIS52), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N2076PECY7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N2076PECY7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N2076PECY7 Supplier Novus Biologicals Supplier No. NBP200076PECY7

Mouse Monoclonal Antibody

Pan-Endothelium Marker Monoclonal specifically detects Pan-Endothelium Marker in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Pan-Endothelium Marker
Applications Flow Cytometry
Classification Monoclonal
Clone HIS52
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen The immunogen for this antibody is Pan endothelium.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.