Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
pan-keratin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | pan-keratin |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5329
![]()
|
Proteintech Group Inc
264111AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is a pan-keratin antibody.
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.Specifications
| pan-keratin | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P02533, P02538, P04264, P05787, P08727, P08729, P08779, P12035, P13645, P13647, P19012, P19013, P35908 | |
| 3848, 3849, 3850, 3851, 3852, 3853, 3855, 3856, 3858, 3861, 3866, 3868, 3880 | |
| KRT5 Fusion Protein Ag24184 | |
| Primary | |
| -20°C | |
| KRT10 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| 58 kDa cytokeratin, CK 5, CK5, Cytokeratin 5, K5, keratin 5, KRT5, KRT5A, Type II keratin Kb5 | |
| KRT1, KRT10, KRT14, KRT15, KRT16, KRT19, KRT2, KRT3, KRT4, KRT5, KRT6A, KRT7, KRT8 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title