Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pan-PAX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | Pan-PAX |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3397
![]()
|
Proteintech Group Inc
213831AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is a Pan-PAX antibody.
This gene encodes a member of the paired box (PAX) family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| Pan-PAX | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O43316, O88436, P09084, P15863, P23759, P23760, P24610, P26367, P32114, P32115, P47239, P47242, P51974, P55771, P63015, P63016, Q00288, Q02548, Q02650, Q02962, Q06710, Q2L4T2 | |
| 114502, 18503, 18504, 18505, 18506, 18507, 18508, 18509, 18510, 18511, 25509, 293992, 311505, 362741, 500453, 500574, 5075, 5076, 5077, 5078, 5079, 5080, 5081, 5083, 7849, 81819, 83630 | |
| Pan-PAX Fusion Protein Ag16018 | |
| Primary | |
| -20°C | |
| PAX1 |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| paired box, paired box 8, Paired box protein Pax, Paired box protein Pax 8, Pan-PAX, PAX 8, PAX8 | |
| Pax1, Pax2, PAX3, PAX4, PAX5, PAX6, PAX7, Pax8, PAX9 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title