Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan PKC, p Thr410 Antibody (HL1278) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295013 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295013 Supplier Novus Biologicals Supplier No. NBP325604
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

pan PKC Monoclonal antibody specifically detects pan PKC in Human, Mouse, Rat, Drosophila samples. It is validated for Western Blot

Specifications

Antigen pan PKC
Applications Western Blot
Classification Monoclonal
Clone HL1278
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS
Gene Alias AAG6,PKCA,PKCalpha,PKC-alpha,PKCI+/-,PRKACA,PRKCA,Protein kinase C,Protein kinase C alpha type
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide surrounding phospho Thr410 of human PKC zeta. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5578
Target Species Human, Mouse, Rat, Drosophila
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.