Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan RhO/RAC/CDC42 Antibody (3L74), Novus Biologicals™
SDP

Catalog No. NBP229740 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229740 0.1 mL
NB019447 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229740 Supplier Novus Biologicals Supplier No. NBP229740
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

pan RhO/RAC/CDC42 Monoclonal specifically detects pan RhO/RAC/CDC42 in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen pan RhO/RAC/CDC42
Applications Western Blot
Classification Monoclonal
Clone 3L74
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation No buffer with No Preservative
Gene Accession No. P61586
Gene Alias h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA
Gene Symbols RHOA
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 73-104 of human Rho (-A, -B, -C), with two terminal lysines added to increase peptide solubility.
Molecular Weight of Antigen 22 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline mTOR Pathway, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 387
Test Specificity Recognizes Rho (-A, -B, -C).
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.