Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
pan RhO/RAC/CDC42 Antibody (3L74), Novus Biologicals™

Rabbit Monoclonal Antibody
$181.90 - $731.50
Specifications
Antigen | pan RhO/RAC/CDC42 |
---|---|
Clone | 3L74 |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
pan RhO/RAC/CDC42 Monoclonal specifically detects pan RhO/RAC/CDC42 in Human, Rat samples. It is validated for Western Blot.Specifications
pan RhO/RAC/CDC42 | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Rabbit | |
mTOR Pathway, Signal Transduction, Wnt Signaling Pathway | |
No buffer with No Preservative | |
h12, oncogene RHO H12, ras homolog gene family, member A, Rho cDNA clone 12, Rho12, RhoA, RHOH12ARH12ARHAAplysia ras-related homolog 12, small GTP binding protein RhoA, transforming protein RhoA | |
RHOA | |
IgG | |
Affinity Purified | |
Recognizes Rho (-A, -B, -C). |
3L74 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
P61586 | |
387 | |
KLH-conjugated, synthetic peptide corresponding to amino acids 73-104 of human Rho (-A, -B, -C), with two terminal lysines added to increase peptide solubility. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
22 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title