Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan-Ubiquitin Remnant Motif Rabbit, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167085 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167085 100 μg
NB167086 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167085 Supplier Novus Biologicals Supplier No. NBP315995100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

pan-Ubiquitin Remnant Motif Polyclonal antibody specifically detects pan-Ubiquitin Remnant Motif in All species samples. It is validated for Western Blot

Specifications

Antigen pan-Ubiquitin Remnant Motif
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence containing ubiquitinated Motif (K-Epsilon-GG)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.