Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP244286 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244286 Supplier Novus Biologicals Supplier No. NBP244286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects PAN3 in Human samples, and it is validated for Immunoprecipitation, Western Blotting

PAN3 Polyclonal specifically detects PAN3 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PAN3
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias hPan3, PAB-dependent poly(A)-specific ribonuclease subunit 3, PABP1-dependent poly A-specific ribonuclease subunit PAN3, PABP-dependent poly(A) nuclease 3, PAN3 poly(A) specific ribonuclease subunit homolog (S. cerevisiae), TS
Gene Symbols PAN3
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residue 175 to 225 of human PAB-dependent poly using the numbering given in entry Q58A45.3 (GeneID 255967).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 255967
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.