Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic Amylase Alpha Antibody (OTI1E3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP273239B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273239B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273239B Supplier Novus Biologicals Supplier No. NBP273239B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pancreatic Amylase Alpha Monoclonal specifically detects Pancreatic Amylase Alpha in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Pancreatic Amylase Alpha
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E3
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase
Gene Symbols AMY2A
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 135-408 of human Pancreatic Amylase Alpha (NP_000690) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 279.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.