Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic Amylase Alpha Mouse anti-Rat, Clone: RPA-B5, Novus Biologicals™
SDP

Catalog No. NBP150750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150750 0.1 mg
NB018666 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP150750 Supplier Novus Biologicals Supplier No. NBP150750

Mouse Monoclonal Antibody

Pancreatic Amylase Alpha Monoclonal specifically detects Pancreatic Amylase Alpha in Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Pancreatic Amylase Alpha
Applications Western Blot, ELISA
Classification Monoclonal
Clone RPA-B5
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA
Gene Accession No. P00689
Gene Alias AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase
Gene Symbols AMY2A
Host Species Mouse
Immunogen Recombinant protein expressed in E. coli
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 279
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.