Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pancreatic Amylase Alpha Mouse anti-Rat, Clone: RPA-B5, Novus Biologicals™

Mouse Monoclonal Antibody
$220.00 - $524.00
Specifications
Antigen | Pancreatic Amylase Alpha |
---|---|
Clone | RPA-B5 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot, ELISA |
Applications | Western Blot, ELISA |
Description
Pancreatic Amylase Alpha Monoclonal specifically detects Pancreatic Amylase Alpha in Rat samples. It is validated for Western Blot, ELISA.Specifications
Pancreatic Amylase Alpha | |
1.0 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
P00689 | |
279 | |
Recombinant protein expressed in E. coli | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
RPA-B5 | |
Western Blot, ELISA | |
Monoclonal | |
Mouse | |
Rat | |
AMY2, AMY2A amylase, alpha 2A (pancreatic), AMY2B, PA, pancreatic alpha-amylase | |
AMY2A | |
IgG1 | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title