Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic Islet Lysate Antibody, Novus Biologicals™
SDP

Catalog No. NBP244126 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244126 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244126 Supplier Novus Biologicals Supplier No. NBP244126
Add to Cart
Add to Cart

Canine Polyclonal Antibody

Specifically detects Pancreatic Islet Lysate in Porcine samples, and it is validated for Sandwich ELISA, Western Blotting

Pancreatic Islet Lysate Polyclonal specifically detects Pancreatic Islet Lysate in Porcine samples. It is validated for Western Blot, Sandwich ELISA.

Specifications

Antigen Pancreatic Islet Lysate
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Sandwich ELISA 1:50 -1:200
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P01300
Host Species Canine
Immunogen Sonicated crude membrane fraction of porcine pancreatic islets of langerhans cells (Uniprot: P01300)
Molecular Weight of Antigen 7.29 kDa
Purification Method Multi-step
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.