Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic trypsin inhibitor Antibody (18), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP22388A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22388A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22388A32 Supplier Novus Biologicals Supplier No. NBP223588AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pancreatic trypsin inhibitor Monoclonal specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Pancreatic trypsin inhibitor
Applications Western Blot, ELISA
Classification Monoclonal
Clone 18
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor
Gene Symbols PTI
Host Species Mouse
Immunogen Aprotinin from bovine lung adsorbed onto aluminum hydroxide gel
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 404172
Test Specificity Clone 18 binds bovine aprotinin immobilized on a solid phase or free in solution.
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.