Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic trypsin inhibitor Antibody (21) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019348 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019348 0.025 mL
NBP22358901 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019348 Supplier Novus Biologicals Supplier No. NBP2235890.025ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pancreatic trypsin inhibitor Monoclonal specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Pancreatic trypsin inhibitor
Applications ELISA, Western Blot
Classification Monoclonal
Clone 21
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor
Host Species Mouse
Immunogen Aprotinin from bovine lung adsorbed onto aluminum hydroxide gel
Purification Method Protein A or G purified
Quantity 0.025 mL
Primary or Secondary Primary
Gene ID (Entrez) 404172
Test Specificity Clone 21 binds bovine aprotinin immobilized on a solid phase or free in solution.
Target Species Bovine
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.