Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pancreatic trypsin inhibitor Antibody (35), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB355615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355615 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355615 Supplier Novus Biologicals Supplier No. NBP223590MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pancreatic trypsin inhibitor Monoclonal antibody specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA

Specifications

Antigen Pancreatic trypsin inhibitor
Applications Western Blot, ELISA
Classification Monoclonal
Clone 35
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor
Host Species Mouse
Immunogen Aprotinin from bovine lung
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404172
Target Species Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.