Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Panendothelial Cell Antigen Antibody (MECA-32) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10077668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10077668 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10077668 Supplier Novus Biologicals Supplier No. NB10077668
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Panendothelial Cell Antigen Monoclonal specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Panendothelial Cell Antigen
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MECA-32
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Flow Cytometry <= 0.25 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200 - 1:500. Use reported in scientific literature (PMID 26586691), Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
Gene Accession No. Q91VC4
Gene Alias MECA32, MECA-32, Panendothelial Cell Antigen
Gene Symbols Plvap
Host Species Rat
Immunogen Mouse lymph node stromal cells.
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84094
Test Specificity Pan endothelial cells except for CNS & muscle capillaries.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.