Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PANK2 Antibody (OTI3H9), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP273241G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273241G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273241G Supplier Novus Biologicals Supplier No. NBP273241G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PANK2 Monoclonal specifically detects PANK2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PANK2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3H9
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C20orf48, EC 2.7.1.33, FLJ17232, Hallervorden-Spatz syndrome, HARPFLJ11729, hPanK2, HSS, MGC15053, NBIA1, pantothenate kinase 2, pantothenate kinase 2, mitochondrial, Pantothenic acid kinase 2, PKANneurodegeneration with brain iron accumulation 1 (Hallervorden-Spatz syndrome)
Gene Symbols PANK2
Host Species Mouse
Immunogen Full length recombinant protein of human PANK2 (NP_705902) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80025.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.