Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pannexin-1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB341472 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341472 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341472 Supplier Novus Biologicals Supplier No. NBP282019PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pannexin-1 Polyclonal antibody specifically detects Pannexin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Pannexin-1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias MGC21309, MRS1innexin, pannexin 1, pannexin-1, PX1, UNQ2529
Host Species Rabbit
Immunogen Rabbit polyclonal Pannexin-1 antibody was raised against a 17 amino acid peptide near the amino terminus of human Pannexin-1. The immunogen is located within amino acids 70 - 120 of Pannexin-1. Amino Acid Squence: CAVQQKNSLQSESGNLPL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 24145
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.