Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Papain Antibody, Novus Biologicals™
SDP

Catalog No. NBP244229 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244229 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244229 Supplier Novus Biologicals Supplier No. NBP244229
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects Papain in Plant samples, and it is validated for ELISA, Immunohistochemistry, Western Blotting

Papain Polyclonal specifically detects Papain in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Papain
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:10000-1:50000, Immunohistochemistry 1:500-1:3000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P00784
Gene Alias Allergen Car p 1, Papaya proteinase I, PPI
Host Species Goat
Immunogen Papain Antibody was produced by repeated immunizations with pork Papain protein isolated from papaya. (Uniprot: P00784)
Molecular Weight of Antigen 38.92 kDa
Purification Method Multi-step
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Plant
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.