Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAPD1 Mouse anti-Human, DyLight 650, Clone: 3D2, Novus Biologicals™
SDP

Catalog No. NBP243679C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP243679C 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP243679C Supplier Novus Biologicals Supplier No. NBP243679C

Mouse Monoclonal Antibody

Specifications

Antigen PAPD1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3D2
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.7.19, FLJ10486, mitochondrial poly(A) polymerase, mtPAP, PAP, PAP associated domain containing 1, PAP-associated domain-containing protein 1, PAPD1SPAX4, poly(A) RNA polymerase, mitochondrial, Polynucleotide adenylyltransferase, Terminal uridylyltransferase 1, TUTase 1
Gene Symbols MTPAP
Host Species Mouse
Immunogen The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 55149
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.