Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAPD5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP262796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262796 Supplier Novus Biologicals Supplier No. NBP262796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PAPD5 Polyclonal specifically detects PAPD5 in Human samples. It is validated for Western Blot.

Specifications

Antigen PAPD5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation Whole antiserum with 0.05% Sodium Azide
Gene Alias EC 2.7.7, EC 2.7.7.-, FLJ40270, PAP associated domain containing 5, Terminal uridylyltransferase 3, Topoisomerase-related function protein 4-2, TRF4-2PAP-associated domain-containing protein 5, TUTase 3
Gene Symbols TENT4B
Host Species Rabbit
Immunogen Human PAPD5 (PAP associated domain containing 5)
Purification Method Whole Antiserum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 64282
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.