Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Papilin Antibody (5F2D10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP237321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237321 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237321 Supplier Novus Biologicals Supplier No. NBP237321
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Papilin Monoclonal specifically detects Papilin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Papilin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5F2D10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias papilin, Papilin, Proteoglycan-Like Sulfated Glycoprotein, PAPLN
Gene Symbols PAPLN
Host Species Mouse
Immunogen Purified recombinant fragment of human PAPLN (AA: 766-870) expressed in E. Coli.
Molecular Weight of Antigen 137.7 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89932
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.