Learn More
Johannes Lieder Papilionaceous Plants (Fabaceae)
Shop All Johannes Lieder GmbH & Co. KG Products
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Description
- 6130 Fabaceae (Papilionaceae)
- 6 Microscope Slides
- Pea (Pisum sativum), stem and petiolar tendril t.s.
- Pea, root with root nodules and nitrogen fixing bacteria t.s.
- Bean (Phaseolus vulgaris), flower bud t.s.
- Bean, flower bud l.s.
- Bean, flower showing young fruit l.s.
- Bean, pod with pericarp and seed t.s.
Specifications
Specifications
Classification | Botany |
Material | Elastin Stained Glass |
Mounting Type | Cross and Longitudinal Section |
Type | Microscope Slides |
Slide Type | Prepared Slide |