Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pappalysin-1/PAPP-A Antibody (10E1cc), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB359486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359486 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359486 Supplier Novus Biologicals Supplier No. NB1208346AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pappalysin-1/PAPP-A Monoclonal antibody specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for Western Blot, Immunoprecipitation, ELISA

Specifications

Antigen Pappalysin-1/PAPP-A
Applications Western Blot, Immunoprecipitation, ELISA
Classification Monoclonal
Clone 10E1cc
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunoprecipitation, Sandwich ELISA
Formulation 50mM Sodium Borate
Gene Alias ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified human PAPP-A/proMBP (heterotetrameric PAPP-A, htPAPP-A) complex.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5069
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.