Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pappalysin-1/PAPP-A Antibody (7A6), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB10073060C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073060C 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073060C Supplier Novus Biologicals Supplier No. NB10073060C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pappalysin-1/PAPP-A Monoclonal specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for ELISA, Immunoprecipitation, Sandwich ELISA.

Specifications

Antigen Pappalysin-1/PAPP-A
Applications ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 7A6
Conjugate DyLight 650
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1
Gene Symbols PAPPA
Host Species Mouse
Immunogen purified human PAPP-A/proMBP (heterotetrameric PAPP-A, htPAPP-A) complex.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5069
Test Specificity Human PAPP-A antigen purified from placental blood (heterotetrameric complex, consisting of PAPP-A subunits and pro-MBP subunits).
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.