Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAR/pADPr Antibody (10H), Novus Biologicals™
SDP

Catalog No. NB036630 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036630 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036630 Supplier Novus Biologicals Supplier No. NBP289039
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PAR/pADPr Monoclonal specifically detects PAR/pADPr in Human, Mouse, Rat, Drosophila samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PAR/pADPr
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10H
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5-20 ug/ml, Immunohistochemistry-Paraffin 5-20 ug/ml
Formulation 50mM HEPES, pH 7.4, containing 100mM sodium chloride, 1% BSA
Gene Alias Poly(ADP-ribose)
Host Species Mouse
Immunogen Purified poly(ADP-ribose).
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Drosophila
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.